Diethyl malonate, 99+%, Thermo Scientific Chemicals
Diethyl malonate, 99+%, Thermo Scientific Chemicals
Diethyl malonate, 99+%, Thermo Scientific Chemicals
Diethyl malonate, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyl malonate, 99+%, Thermo Scientific Chemicals
CAS: 105-53-3 | C7H12O4 | 160.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR114450250
Price (MYR)
374.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
374.00
Add to cart
Chemical Identifiers
CAS9004-57-3
IUPAC Name2-[butyl({4-[2-(2,6-dicyano-4-nitrophenyl)diazen-1-yl]-3-methylphenyl})amino]ethyl acetate
Molecular FormulaC23H24N6O4
InChI KeyARSKJXYLLONUAJ-UHFFFAOYSA-N
SMILESCCCCN(CCOC(C)=O)C1=CC(C)=C(C=C1)N=NC1=C(C=C(C=C1C#N)[N+]([O-])=O)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Viscosity8 to 12 cP (5% in 80/20 toluene/ethanol, 25°C)
Appearance (Form)Powder or granules
Assay47.5 to 49.5 % (Ethoxyl content)
Appearance (Color)White to yellow