Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyldithiocarbamic acid, silver salt, 99%, for analysis, Thermo Scientific Chemicals
CAS: 1470-61-7 | C5H10AgNS2 | 256.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR114291000
Price (MYR)
7,216.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
7,216.00
Add to cart
Specifications
Chemical Name or MaterialDiethyldithiocarbamic acid, silver salt
Name NoteFor analysis
CAS1470-61-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
View more
For analysis
RUO – Research Use Only