Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals
Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals
Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals
Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals
CAS: 1115-30-6 | C10H16O5 | 216.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR113951000
Price (MYR)
1,431.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,431.00
Add to cart
Chemical Identifiers
CAS12036-02-1
IUPAC Namedioxoosmium
Molecular FormulaO2Os
InChI KeyXSXHWVKGUXMUQE-UHFFFAOYSA-N
SMILESO=[Os]=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (unspecified)≥83.0% Osmium
Appearance (Color)Black to black-brown to grey