Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals
Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals
Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals
Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Diethyl acetylsuccinate, 99%, Thermo Scientific Chemicals
CAS: 1115-30-6 | C10H16O5 | 216.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR113950250
Price (MYR)
559.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
559.00
Add to cart
Chemical Identifiers
CAS582-25-2
IUPAC Namepotassium benzoate
Molecular FormulaC7H5KO2
InChI KeyXAEFZNCEHLXOMS-UHFFFAOYSA-M
SMILES[K+].[O-]C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Formpowder
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)>98.5%