N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals
N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals
N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals
N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N,N'-Dicyclohexylcarbodiimide, 99%, Thermo Scientific Chemicals
N,N-Dicyclohexylcarbodiimide, 99%, C13H22N2, CAS Number-538-75-0, carbodiimide, dicyclohexyl, bis cyclohexyl carbodiimide, dcc, 1,3-dicyclohexylcarbodiimide, carbodicyclohexylimide, n,n'-dicyclohexylcarbodiimide, n,n-dicyclohexylcarbodiimide, dcci, dicyclohexylcarbodiimide, dccd, 1kg
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
100 g
1 kg
5 kg
25 kg
Packaging:
View all
Glass bottle
Catalog number ACR113902500
Price (MYR)
1,118.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,118.00
Add to cart
Chemical Identifiers
CAS304-88-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light beige
Appearance (Form)Crystalline powder or crystals or needles
Infrared spectrumConforms
Melting point119°C to 123°C
HPLC>=97.5 %
Coupling reagent
RUO – Research Use Only