2,4-Dichlorophenoxyacetic acid, 97%, Thermo Scientific Chemicals
2,4-Dichlorophenoxyacetic acid, 97%, Thermo Scientific Chemicals
2,4-Dichlorophenoxyacetic acid, 97%, Thermo Scientific Chemicals
2,4-Dichlorophenoxyacetic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2,4-Dichlorophenoxyacetic acid, 97%, Thermo Scientific Chemicals
CAS: 94-75-7 | C8H6Cl2O3 | 221.033 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Catalog number ACR113631000
Price (MYR)
450.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
450.00
Add to cart
Specifications
Chemical Name or Material2, 4-Dichlorophenoxyacetic acid
CAS94-75-7
CAS Min %99.0
CAS Max %100.0
Health Hazard 2GHS H Statement:
May cause respiratory irritation.
Harmful to aquatic life with long lasting effects.