Dichloromethane, 99.8+%, for analysis, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.8+%, for analysis, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.8+%, for analysis, stabilized with amylene, Thermo Scientific Chemicals
Dichloromethane, 99.8+%, for analysis, stabilized with amylene, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dichloromethane, 99.8+%, for analysis, stabilized with amylene, Thermo Scientific Chemicals
CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number ACR113460010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDichloromethane
Melting Point-97.0°C
CAS75-09-2
Health Hazard 1Warning
Health Hazard 2GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. May cause drowsiness or dizziness. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure.
View more
For analysis
RUO – Research Use Only