Dibromomethane, 99%, Thermo Scientific Chemicals
Dibromomethane, 99%, Thermo Scientific Chemicals
Dibromomethane, 99%, Thermo Scientific Chemicals
Dibromomethane, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dibromomethane, 99%, Thermo Scientific Chemicals
CAS: 74-95-3 | CH2Br2 | 173.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number ACR112831000
Price (MYR)
391.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
391.00
Add to cart
Chemical Identifiers
CAS694-83-7
IUPAC Name(1R,2S)-cyclohexane-1,2-bis(aminium)
Molecular FormulaC6H16N2
InChI KeySSJXIUAHEKJCMH-OLQVQODUSA-P
SMILES[NH3+][C@H]1CCCC[C@H]1[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<75 APHA
Infrared spectrumConforms
GC>=98.5 %
Refractive index1.4860 to 1.4920 (20°C, 589 nm)
Appearance (Form)Clear liquid