Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
CAS: 120-46-7 | C15H12O2 | 224.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
Packaging:
View all
Plastic bottle
Catalog number ACR112605000
Price (MYR)
3,420.00
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
3,420.00
Add to cart
Chemical Identifiers
CAS123-91-1
IUPAC Name1,4-dioxane
Molecular FormulaC4H8O2
InChI KeyRYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILESC1COCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤0.05%
StabilizerIf present, report quantity and identity
Appearance (Color)Colorless to pale yellow
FormLiquid
Color (APHA)≤20