Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Dibenzoylmethane, 98%, Thermo Scientific Chemicals
CAS: 120-46-7 | C15H12O2 | 224.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
500 g
Packaging:
View all
Glass Bottle
Catalog number ACR112600100
Price (MYR)
380.00
Add to cart
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
380.00
Add to cart
Specifications
Chemical Name or MaterialDibenzoylmethane
Name Note98%
CAS120-46-7
Melting Point75°C to 79°C
ColorYellow