3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals
3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals
3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals
3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals
CAS: 619-05-6 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number ACR112101000
Price (MYR)
1,119.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,119.00
Add to cart
Chemical Identifiers
CAS7758-87-4
IUPAC Nametricalcium diphosphate
Molecular FormulaCa3O8P2
InChI KeyQORWJWZARLRLPR-UHFFFAOYSA-H
SMILES[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Titration Complexometric>=95 %