3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals
3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals
3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals
3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
3,4-Diaminobenzoic acid, 97%, Thermo Scientific Chemicals
CAS: 619-05-6 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number ACR112100250
Price (MYR)
549.00
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
549.00
Add to cart
Chemical Identifiers
CAS17016-83-0
IUPAC Name(4S)-4-(propan-2-yl)-1,3-oxazolidin-2-one
Molecular FormulaC6H11NO2
InChI KeyYBUPWRYTXGAWJX-RXMQYKEDSA-N
SMILESCC(C)[C@H]1COC(=O)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Melting point70°C to 74°C
Specific optical rotation-16° to -20° (20°C, 589 nm) (c=1, ethanol)