Deoxybenzoin, 97%, Thermo Scientific Chemicals
Deoxybenzoin, 97%, Thermo Scientific Chemicals
Deoxybenzoin, 97%, Thermo Scientific Chemicals
Deoxybenzoin, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Deoxybenzoin, 97%, Thermo Scientific Chemicals
CAS: 451-40-1 | C14H12O | 196.25 g/mol
Have Questions?
Catalog NumberQuantity
ACR11196025025 g
Catalog number ACR111960250
Price (MYR)
530.00
EA
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
530.00
EA
Add to cart
Chemical Identifiers
CAS75-75-2
IUPAC Namemethanesulfonic acid
Molecular FormulaCH4O3S
InChI KeyAFVFQIVMOAPDHO-UHFFFAOYSA-N
SMILESCS(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<150 APHA
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Appearance (Form)Clear liquid
Refractive index1.4285 to 1.4315 (20°C, 589 nm)