Decane, 99+%, pure, Thermo Scientific Chemicals
Decane, 99+%, pure, Thermo Scientific Chemicals
Decane, 99+%, pure, Thermo Scientific Chemicals
Decane, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Decane, 99+%, pure, Thermo Scientific Chemicals
Decane, >-99.0%, C10H22, CAS Number-124-18-5, ccris 653, decane, analytical standard, decyl hydride, dsstox_cid_4913, dsstox_gsid_24913, dsstox_rid_77577, hsdb 63, n-decane, nonane, methyl, unii-nk85062oiy, 2.5L, 99+%, 01, 168, CHEBI:41808, Colorless, 0.7350g/mL, 204-686-4, 142.28, Pure, Danger
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Packaging:
View all
Glass Bottle
Catalog number ACR111875000
Price (MYR)
1,084.00
Add to cart
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,084.00
Add to cart
Chemical Identifiers
CAS14855-76-6
IUPAC Namebis[4-(dimethylamino)phenyl][4-(ethyldimethylazaniumyl)phenyl]methylium bromide chloride
Molecular FormulaC27H35BrClN3
InChI KeyIDAQSADEMXDTKN-UHFFFAOYSA-L
SMILES[Cl-].[Br-].CC[N+](C)(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofA0.100% w/v ? 0.005% w/v
Assay from Supplier's CofATransition: pH 0.2 (yellow) - pH 1.8 (blue)