Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Decahydronaphthalene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Packaging:
Glass bottle
Catalog number ACR111845000
Price (MYR)
600.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
600.00
Add to cart
Specifications
Chemical Name or MaterialDecahydronaphthalene
Name NoteMixture of cis and trans
CAS91-17-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Toxic if inhaled.
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.