Cytidine, 99+%
Cytidine, 99+%
Cytidine, 99+%
Thermo Scientific Chemicals
Cytidine, 99+%
CAS: 65-46-3 | C9H13N3O5 | 243.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
Catalog number ACR111812500
Price (MYR)
4,469.00
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
4,469.00
Add to cart
Chemical Identifiers
CAS151-21-3
IUPAC Namesodium dodecyl sulfate
Molecular FormulaC12H25NaO4S
InChI KeyDBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay>=99.0 %
Chloride (Cl)=<1000 ppm
UVat 260 nm A: =<0.05