Cytidine, 99+%
Cytidine, 99+%
Cytidine, 99+%
Thermo Scientific Chemicals
Cytidine, 99+%
CAS: 65-46-3 | C9H13N3O5 | 243.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
250 g
Catalog number ACR111810100
Price (MYR)
566.00
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (MYR)
566.00
Add to cart
Chemical Identifiers
CAS13126-12-0
IUPAC Namerubidium(1+) nitrate
Molecular FormulaNO3Rb
InChI KeyRTHYXYOJKHGZJT-UHFFFAOYSA-N
SMILES[Rb+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤ 1%
Appearance (Color)White
FormCrystalline