Cyclopropanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Cyclopropanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Cyclopropanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Cyclopropanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclopropanecarboxylic acid chloride, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number ACR111630250
Price (MYR)
616.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
616.00
Add to cart
Chemical Identifiers
CAS1878-84-8
IUPAC Name2-(4-hydroxyphenoxy)acetate
Molecular FormulaC8H7O4
InChI KeyPKGWLCZTTHWKIZ-UHFFFAOYSA-M
SMILESOC1=CC=C(OCC([O-])=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥98.0 to ≤102.0%
Assay (Silylated GC)≥98.0%
Melting Point (clear melt)150.0-158.0°C