Cyclopentylamine, 99+%, Thermo Scientific Chemicals
Cyclopentylamine, 99+%, Thermo Scientific Chemicals
Cyclopentylamine, 99+%, Thermo Scientific Chemicals
Cyclopentylamine, 99+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclopentylamine, 99+%, Thermo Scientific Chemicals
CAS: 1003-03-8 | C5H11N | 85.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR111590050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCyclopentylamine
Name Note99+%
CAS1003-03-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Causes serious eye damage.
Harmful if inhaled.
Highly flammable liquid and vapor.