Cyclopentanol, 99%, Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
CAS: 96-41-3 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ACR111525000
Price (MYR)
845.00
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
845.00
Add to cart
Chemical Identifiers
CAS107-05-1
IUPAC Name3-chloroprop-1-ene
Molecular FormulaC3H5Cl
InChI KeyOSDWBNJEKMUWAV-UHFFFAOYSA-N
SMILESClCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4135 to 1.4175 (20°C, 589 nm)