Cyclopentanol, 99%, Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclopentanol, 99%, Thermo Scientific Chemicals
CAS: 96-41-3 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ACR111521000
Price (MYR)
419.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
419.00
Add to cart
Specifications
Chemical Name or MaterialCyclopentanol
Name Note99%
CAS96-41-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Flammable liquid and vapor.