Cyclopentanecarboxylic Acid, 98+%, Thermo Scientific Chemicals
Cyclopentanecarboxylic Acid, 98+%, Thermo Scientific Chemicals
Cyclopentanecarboxylic Acid, 98+%, Thermo Scientific Chemicals
Cyclopentanecarboxylic Acid, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclopentanecarboxylic Acid, 98+%, Thermo Scientific Chemicals
CAS: 3400-45-1 | C6H10O2 | 114.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR111491000
Price (MYR)
2,136.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,136.00
Add to cart
Specifications
Chemical Name or MaterialCyclopentanecarboxylic acid
Name Note98+%
CAS3400-45-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: