Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
CAS: 827-52-1 | C12H16 | 160.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number ACR111295000
Price (MYR)
1,816.00
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,816.00
Add to cart
Chemical Identifiers
CAS176763-62-5
IUPAC Name2,4-di-tert-butyl-6-{[(2-{[(3,5-di-tert-butyl-6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}cyclohexyl)amino]methylidene}cyclohexa-2,4-dien-1-one cobalt
Molecular FormulaC36H54CoN2O2
InChI KeyXJWCTQVLJVAWMC-UHFFFAOYNA-N
SMILES[Co].CC(C)(C)C1=CC(=CNC2CCCCC2NC=C2C=C(C=C(C2=O)C(C)(C)C)C(C)(C)C)C(=O)C(=C1)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red-brown
Infrared spectrumConforms
Elemental Analysis>=97.5 %
Appearance (Form)Powder