Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexylbenzene, 98%, Thermo Scientific Chemicals
CAS: 827-52-1 | C12H16 | 160.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Catalog number ACR111291000
Price (MYR)
613.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
613.00
Add to cart
Chemical Identifiers
CAS827-15-6
IUPAC Name1,2,3,4,5-pentafluoro-6-iodobenzene
Molecular FormulaC6F5I
InChI KeyOPYHNLNYCRZOGY-UHFFFAOYSA-N
SMILESFC1=C(F)C(F)=C(I)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %