Cyclohexene oxide, 98%, Thermo Scientific Chemicals
Cyclohexene oxide, 98%, Thermo Scientific Chemicals
Cyclohexene oxide, 98%, Thermo Scientific Chemicals
Cyclohexene oxide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexene oxide, 98%, Thermo Scientific Chemicals
CAS: 286-20-4 | C6H10O | 98.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number ACR111211000
Price (MYR)
502.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
502.00
Add to cart
Chemical Identifiers
CAS7440-57-5
IUPAC Namegold
Molecular FormulaAu
InChI KeyPCHJSUWPFVWCPO-UHFFFAOYSA-N
SMILES[Au]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.025 mm thick
Appearance (Color)Gold
Total Metal Impurities≤0.015%