Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
CAS: 100-64-1 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number ACR111205000
Price (MYR)
497.00
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
497.00
Add to cart
Chemical Identifiers
CAS2052-49-5
IUPAC Nametetrabutylazanium hydroxide
Molecular FormulaC16H37NO
InChI KeyVDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES[OH-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear solution
Color scale=<150 APHA
Titration with HCl38 to 42 %