Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
CAS: 100-64-1 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number ACR111201000
Price (MYR)
328.00
Add to cart
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
328.00
Add to cart
Chemical Identifiers
CAS60-12-8
IUPAC Name2-phenylethan-1-ol
Molecular FormulaC8H10O
InChI KeyWRMNZCZEMHIOCP-UHFFFAOYSA-N
SMILESOCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Refractive index1.5300 to 1.5350 (20°C, 589 nm)
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=98.5 %