Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexanone Oxime 97%, Thermo Scientific Chemicals
CAS: 100-64-1 | C6H11NO | 113.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Plastic Bottle
Catalog number ACR111200025
Price (MYR)
1,278.00
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,278.00
Add to cart
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤ 1.0%
Surface Area100-200m²/g
Appearance (Color)White
Particle Size20nm Average