1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
1,3-Cyclohexanedione, 97%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number ACR111171000
Price (MYR)
565.00
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
565.00
Add to cart
Chemical Identifiers
CAS2456-81-7
IUPAC Name4-(pyrrolidin-1-yl)pyridin-1-ium
Molecular FormulaC9H13N2
InChI KeyRGUKYNXWOWSRET-UHFFFAOYSA-O
SMILESC1CCN(C1)C1=CC=[NH+]C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale yellow
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
FormCrystals or powder or crystalline powder or lumps or fused solid
Assay (GC)≥97.5%
Melting Point (clear melt)52.0-60.0?C