Cyclohexane, 99+%, pure, Thermo Scientific Chemicals
Cyclohexane, 99+%, pure, Thermo Scientific Chemicals
Cyclohexane, 99+%, pure, Thermo Scientific Chemicals
Cyclohexane, 99+%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyclohexane, 99+%, pure, Thermo Scientific Chemicals
CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number ACR111110010
Price (MYR)
502.00
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
502.00
Add to cart
Specifications
Chemical Name or MaterialCyclohexane
Melting Point6.5°C
CAS110-82-7
Health Hazard 1Danger
Health Hazard 2GHS P Statement May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. Causes skin irritation. Very toxic to aquatic life with long lasting effects. Highly flammable liquid and vapor.