Cyanuric chloride, 99%, Thermo Scientific Chemicals
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number ACR110892500
Price (MYR)
435.00
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
435.00
Add to cart
Specifications
Chemical Name or MaterialCyanuric chloride
Name Note0.99
CAS108-77-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
May cause respiratory irritation.
Fatal if inhaled.
Harmful if swallowed.
Reacts violently with water.
View more
C-X bond formation reagent
RUO – Research Use Only