Cyanuric chloride, 99%, Thermo Scientific Chemicals
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Packaging:
View all
Plastic drum
Catalog number ACR110890051
Price (MYR)
2,002.00
Add to cart
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Price (MYR)
2,002.00
Add to cart
Chemical Identifiers
CAS7727-54-0
IUPAC Namediammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaH8N2O8S2
InChI KeyROOXNKNUYICQNP-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals
Titration with KMnO4>=98.0 %
Heavy metals (as Pb)=<0.005 %
Residue after ignition=<0.05 %
View more
C-X bond formation reagent
RUO – Research Use Only