Cyanuric chloride, 99%, Thermo Scientific Chemicals
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cyanuric chloride, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
5 kg
Packaging:
View all
Glass bottle
Catalog number ACR110890010
Price (MYR)
677.00
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
677.00
Add to cart
Chemical Identifiers
CAS11128-95-3
MDL NumberMFCD00145822
SpecificationsSpecification SheetSpecification Sheet
Uniformity coefficient=<1.20
Total volume capacity>=1.10 eq/l (OH) as is
Appearance (Form)Spherical beads
Moisture holding capacity54.0 to 60.0 % (OH)
Harmonic mean size0.580 to 0.680 mm
View more
C-X bond formation reagent
RUO – Research Use Only