4-Cyanopyridine, 98%, Thermo Scientific Chemicals
4-Cyanopyridine, 98%, Thermo Scientific Chemicals
4-Cyanopyridine, 98%, Thermo Scientific Chemicals
4-Cyanopyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Cyanopyridine, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
View all
Plastic bottle
Catalog number ACR110871000
Price (MYR)
444.00
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
444.00
Add to cart
Specifications
Chemical Name or Material4-Cyanopyridine
Name Note98%
CAS100-48-1
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning