4-Cyanopyridine, 98%, Thermo Scientific Chemicals
4-Cyanopyridine, 98%, Thermo Scientific Chemicals
4-Cyanopyridine, 98%, Thermo Scientific Chemicals
4-Cyanopyridine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Cyanopyridine, 98%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number ACR110870250
Price (MYR)
367.00
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
367.00
Add to cart
Chemical Identifiers
CAS634-47-9
IUPAC Name2-chloro-4-methylquinoline
Molecular FormulaC10H8ClN
InChI KeyPFEIMKNQOIFKSW-UHFFFAOYSA-N
SMILESCC1=C2C=CC=CC2=NC(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)53.0-59.0?C
Appearance (Color)White
Assay (GC)≥98.5%