2-Cyanopyridine, 99%, Thermo Scientific Chemicals
2-Cyanopyridine, 99%, Thermo Scientific Chemicals
2-Cyanopyridine, 99%, Thermo Scientific Chemicals
2-Cyanopyridine, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
2-Cyanopyridine, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number ACR110851000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS19767-45-4
IUPAC Namesodium 2-sulfanylethane-1-sulfonate
Molecular FormulaC2H5NaO3S2
InChI KeyXOGTZOOQQBDUSI-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)CCS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Loss on drying=<2 %
Appearance (Color)White to light yellow
Infrared spectrumConforms
Titration Iodometric>=97.5 to 102 % (On dried substance)