Copper(II) acetylacetonate, 98%, Thermo Scientific Chemicals
Copper(II) acetylacetonate, 98%, Thermo Scientific Chemicals
Copper(II) acetylacetonate, 98%, Thermo Scientific Chemicals
Copper(II) acetylacetonate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Copper(II) acetylacetonate, 98%, Thermo Scientific Chemicals
CAS: 13395-16-9 | C10H14CuO4 | 261.76 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number ACR110651000
Price (MYR)
656.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
656.00
Add to cart
Chemical Identifiers
CAS10034-96-5
IUPAC Namemanganese(2+) hydrate sulfate
Molecular FormulaH2MnO5S
InChI KeyISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILESO.[Mn++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale pink to pink
FormCrystals or powder or crystalline powder
Complexometric Titration≥96.0 to ≤104.0%