Cumene, 98%, pure, Thermo Scientific Chemicals
Cumene, 98%, pure, Thermo Scientific Chemicals
Cumene, 98%, pure, Thermo Scientific Chemicals
Cumene, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cumene, 98%, pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass bottle
Catalog number ACR110630025
Price (MYR)
888.00
Add to cart
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
888.00
Add to cart
Chemical Identifiers
CAS7487-94-7
IUPAC Namemercury(2+) dichloride
Molecular FormulaCl2Hg
InChI KeyLWJROJCJINYWOX-UHFFFAOYSA-L
SMILES[Cl-].[Cl-].[Hg++]
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentFe 0.002%
Assay (unspecified)99.5% Cl₂Hg min
CommentResidue after reduction 0.02%
CommentSolution in ethyl ether P.T.