Cumene, 98%, pure, Thermo Scientific Chemicals
Cumene, 98%, pure, Thermo Scientific Chemicals
Cumene, 98%, pure, Thermo Scientific Chemicals
Cumene, 98%, pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cumene, 98%, pure, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Catalog number ACR110630010
Price (MYR)
523.00
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
523.00
Add to cart
Chemical Identifiers
CAS1569-44-4
IUPAC Name3-methylhex-5-en-3-ol
Molecular FormulaC7H14O
InChI KeyGBARKLJMQRUBKV-UHFFFAOYNA-N
SMILESCCC(C)(O)CC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.4355-1.4395 @ 20?C