Citronellol, 95%, Thermo Scientific Chemicals
Citronellol, 95%, Thermo Scientific Chemicals
Citronellol, 95%, Thermo Scientific Chemicals
Citronellol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citronellol, 95%, Thermo Scientific Chemicals
CAS: 106-22-9 | C10H20O | 156.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number ACR110461000
Price (MYR)
434.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
434.00
Add to cart
Chemical Identifiers
CAS109-43-3
IUPAC Name1,10-dibutyl decanedioate
Molecular FormulaC18H34O4
InChI KeyPYGXAGIECVVIOZ-UHFFFAOYSA-N
SMILESCCCCOC(=O)CCCCCCCCC(=O)OCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<40 APHA
Infrared spectrumConforms
Refractive index1.4400 to 1.4420 (20°C, 589 nm)
Appearance (Form)Clear liquid
Water=<0.1 % (K.F.)