Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
CAS: 5392-40-5 | C10H16O | 152.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
5 mL
500 mL
Packaging:
Glass bottle
Catalog number ACR110441000
Price (MYR)
447.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
447.00
Add to cart
Chemical Identifiers
CAS10039-56-2
IUPAC Namesodium hydrate phosphinate
Molecular FormulaH4NaO3P
InChI KeyPLZNPHDJGFDNRM-UHFFFAOYSA-M
SMILESO.[Na+].[O-][PH2]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration99.0 to 103.0 %
Arsenic (As)=<2 ppm
Iron (Fe)=<5 ppm
Calcium (Ca)=<200 ppm