Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Citral, 95%, mixture of cis and trans, Thermo Scientific Chemicals
CAS: 5392-40-5 | C10H16O | 152.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number ACR110440050
Price (MYR)
360.00
Add to cart
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
360.00
Add to cart
Chemical Identifiers
CAS143900-43-0
IUPAC Nametert-butyl 3-hydroxypiperidine-1-carboxylate
Molecular FormulaC10H19NO3
InChI KeyUIJXHKXIOCDSEB-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCCC(O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormSolid
Assay (unspecified)≥96.0%