Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
CAS: 4392-24-9 | C9H9Br | 197.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number ACR110401000
Price (MYR)
2,281.00
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,281.00
Add to cart
Chemical Identifiers
CAS1113-41-3
IUPAC Name2-amino-3-methyl-3-sulfanylbutanoic acid
Molecular FormulaC5H11NO2S
InChI KeyVVNCNSJFMMFHPL-UHFFFAOYNA-N
SMILESCC(C)(S)C(N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with HClO4>=97.5 %
Solubility(5 % in water) Clear colorless