Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
CAS: 4392-24-9 | C9H9Br | 197.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Packaging:
Glass bottle
Catalog number ACR110400250
Price (MYR)
863.00
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
863.00
Add to cart
Chemical Identifiers
CAS2537-48-6
IUPAC Namediethyl (cyanomethyl)phosphonate
Molecular FormulaC6H12NO3P
InChI KeyKWMBADTWRIGGGG-UHFFFAOYSA-N
SMILESCCOP(=O)(CC#N)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
GC>=95 %
Appearance (Form)Liquid
Refractive index1.4312 to 1.4332 (20°C, 589 nm)