Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cinnamyl bromide, 97%, predominantly trans, Thermo Scientific Chemicals
CAS: 4392-24-9 | C9H9Br | 197.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number ACR110400050
Price (MYR)
490.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
490.00
Add to cart
Chemical Identifiers
CAS10378-47-9
IUPAC Nameλ⁴-cerium(4+) tetraamine dihydrate tetrahydrogen sulfate
Molecular FormulaCeH20N4O18S4
InChI KeyVCNAMBGKEDPVGQ-UHFFFAOYSA-J
SMILESN.N.N.N.O.O.[Ce+4].OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O.OS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<0.01 %
Appearance (Color)Yellow to orange
Appearance (Form)Crystalline powder
Titration>=94 %
Insoluble matter=<0.05 % (in dilute sulfuric acid)