Cinnamoyl chloride, 98%, predominantly trans, Thermo Scientific Chemicals
Cinnamoyl chloride, 98%, predominantly trans, Thermo Scientific Chemicals
Cinnamoyl chloride, 98%, predominantly trans, Thermo Scientific Chemicals
Cinnamoyl chloride, 98%, predominantly trans, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Cinnamoyl chloride, 98%, predominantly trans, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number ACR110390050
Price (MYR)
461.00
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
461.00
Add to cart
Specifications
Chemical Name or MaterialCinnamoyl chloride
Name Note98%
CAS102-92-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Harmful if inhaled.
Reacts violently with water.
Contact with water liberates toxic gas.