trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
CAS: 14371-10-9 | C9H8O | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
50 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number ACR110355000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1984-06-1
IUPAC Namesodium octanoate
Molecular FormulaC8H15NaO2
InChI KeyBYKRNSHANADUFY-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or crystals or crystalline powder
Identification (FTIR)Conforms (non-U.S. specification)
Assay (Non-aqueous acid-base Titration)≥95.0 to ≤105.0% (non-U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
Water Content (Karl Fischer Titration)≤3%