trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
trans-Cinnamaldehyde, 99%, Thermo Scientific Chemicals
CAS: 14371-10-9 | C9H8O | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
50 g
500 g
1 kg
Packaging:
View all
Plastic drum
Catalog number ACR110350050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Specifications
Chemical Name or Materialtrans-Cinnamaldehyde
CAS14371-10-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
May cause an allergic skin reaction.
Health Hazard 3GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.