alpha-Chloro-p-xylene, 98%, Thermo Scientific Chemicals
alpha-Chloro-p-xylene, 98%, Thermo Scientific Chemicals
alpha-Chloro-p-xylene, 98%, Thermo Scientific Chemicals
alpha-Chloro-p-xylene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
alpha-Chloro-p-xylene, 98%, Thermo Scientific Chemicals
CAS: 104-82-5 | C8H9Cl | 140.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
Glass bottle
Catalog number ACR110171000
Price (MYR)
657.00
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
657.00
Add to cart
Specifications
Chemical Name or Materialα-Chloro-p-xylene
Name Note99+%
CAS104-82-5
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger