4-Chlorostyrene, 99%, stabilized, Thermo Scientific Chemicals
4-Chlorostyrene, 99%, stabilized, Thermo Scientific Chemicals
4-Chlorostyrene, 99%, stabilized, Thermo Scientific Chemicals
4-Chlorostyrene, 99%, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Chlorostyrene, 99%, stabilized, Thermo Scientific Chemicals
CAS: 1073-67-2 | C8H7Cl | 138.59 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number ACR110090100
Price (MYR)
620.00
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
620.00
Add to cart
Chemical Identifiers
CAS1314-87-0
IUPAC Nameλ²-lead(2+) sulfanediide
Molecular FormulaPbS
InChI KeyFILXVLQXZYXBSJ-UHFFFAOYSA-N
SMILES[S--].[Pb++]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.005% max.